Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Fetal Clone II PROMO

Manufacturer:  Cytiva 29648031

Catalog No. SH3006HILR1


Add to cart
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Filtered using sequential 100 nm pore-size rated filters.
Suitable for use with a wide variety of cell lines
FetalClone II Serum (U.S.) Heat Inactivated
≤ 10 EU/mL
Typically less than 2 μg/mL
U.S.
No
1 mL
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
FetalClone II
500 mL
Sterile
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
Yes
≤ 20 mg/dL
U.S.
Heat Inactivated
<-5°C
USDA Inspected
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit