Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Fetal Clone II PROMO

Manufacturer:  Cytiva 29647949

Catalog No. SH6603HIRR1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

500 mL
Filtered using sequential 100 nm pore-size rated filters.
FetalClone II Serum (U.S.) Heat Inactivated
≤ 10 EU/mL
Typically less than 2 μg/mL
United States
No
1 mL
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2°C to 8°C.
Fetal Clone II
Suitable for Use with a Wide Variety of Cell Lines
Sterile
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
Yes
≤ 20 mg/dL
United States
Heat Inactivated
<-5°C
USDA Inspected
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit