Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Fetal Clone II

$418.00

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
Endotoxin Concentration ≤ 10 EU/mL
For Use With (Application) Suitable for use with a wide variety of cell lines
Heat Inactivated Yes
Hemoglobin Concentration ≤ 20 mg/dL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Processing Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Processing Price Quantity & Availability  
SH3006603HI
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
SH30066.03HI
500mL Heat Inactivated
Each for $418.00
Add to cart
 
Specifications

Specifications

Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
Suitable for use with a wide variety of cell lines
≤ 20 mg/dL
U.S.
Filtered using sequential 100 nm pore-size rated filters.
USDA Inspected
Fetal Clone II
Sterile
≤ 10 EU/mL
Yes
Typically less than 2 μg/mL
No
<-5°C
U.S.
1mL
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit