Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ FetalClone™ I Serum (U.S.) PROMO

Optimized for growth of hybridomas, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ I Serum, a fetal bovine serum (FBS) alternative containing comparable IgG levels to those found in FBS.

Manufacturer:  Cytiva 29648027

Catalog No. SH300803LR1


Add to cart

Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • Demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • Specifications

    Specifications

    Filtered using sequential 100 nm pore-size rated filters.
    Sterile
    FetalClone I Serum (U.S.)
    ≤ 10 EU/mL
    Typically less than 2 μg/mL
    U.S.
    500 mL
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
    FetalClone I
    500 mL
    Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
    Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
    No
    ≤ 20 mg/dL
    No
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    USDA Inspected
    Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit