Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.) PROMO

HyClone™ FetalClone™ II Serum (U.S.) PROMO is an economic alternative to FBS

Manufacturer:  Cytiva 29490099

Catalog No. SH3006603R0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
Specifications

Specifications

Sterile
FetalClone II Serum (U.S.)
≤ 10 EU/mL
≤ 2μg/mL
U.S.
None
< -5°C
USDA Inspected
Bluetongue
Sequentially with 100 nm pore-size rated filters
Albumin
No
≤ 20 mg/dL
No
500 mL
Once thawed, sera should be stored at 2° to 8°C
FetalClone II
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit