Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.) PROMO

Optimized for the growth of CHO cells and derivatives.

Manufacturer:  Cytiva 29648033

Catalog No. SH300640LR1


Add to cart

Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • Specifications

    Specifications

    Filtered through sequential 100 nm pore-size rated filters
    HyClone™
    Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
    No
    ≤ 20 mg/dL
    No
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    FetalClone II Serum
    500 mL
    Sterile
    ≤ 10 EU/mL
    Typically less than 2 μg/mL
    U.S.
    Irradiated (25 to 40 doses)
    Yes
    Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit