Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.) PROMO

Developed for growth of CHO cells and derivatives, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ II Serum, a fetal bovine serum (FBS) alternative, containing additional growth factors and supplements than basic formulations.

Manufacturer:  Cytiva 29648030

Catalog No. SH300660LR1


Add to cart

Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
Specifications

Specifications

Filtered using sequential 100 nm pore-size rated filters.
500 mL
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
No
≤ 20 mg/dL
No
500 mL
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
FetalClone II
Sterile
FetalClone II Serum (U.S.)
≤ 10 EU/mL
Typically less than 2 μg/mL
U.S.
None
<-5°C
USDA Inspected
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit