Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ FetalClone™ II Serum, US Origin PROMO

Offers the same basic formulation as FetalClone™ I with additional growth factors and supplements. Shows optimized growth of CHO cells and derivatives, with improved cell growth performance relative to fetal bovine serum (FBS).

Manufacturer:  Cytiva 29648032

Catalog No. SH3006IHLR1


Add to cart
This item is not returnable. View return policy

Description

Description

  • A supplement used to enrich cell culture performance
  • Delivers both cost-effectiveness and high performance
  • Commonly used in cell culture and bioprocessing applications
  • Low endotoxin and hemoglobin levels
  • High lot-to-lot consistency
Specifications

Specifications

Filtered through three sequential 100 nm pore size-rated filters
United States
FetalClone™
500 mL
≤ 10 EU/mL
≤ 20 mg/dL
Heat Inactivated, Irradiated
Serum
Frozen
Liquid
Sterile
Manufactured using ISO 9001- and ISO 13485-Certified Processes
Yes
No
Store at or below -10°C
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine Respiratory Syncytial Virus, Bovine Viral Diarrhea Virus, Rabies, and Reovirus
Safety and Handling

Safety and Handling

ShelfLife : 6 Weeks (Thawed)

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit