Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ FetalClone™ III Serum (U.S.) PROMO

HyClone™ FetalClone™ III Serum (U.S.) PROMO is an economic alternative to FBS

Manufacturer:  Cytiva 29490100

Catalog No. SH3010903R0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
Specifications

Specifications

Sterile
Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Albumin
No
≤ 20 mg/dL
No
500 mL
Once thawed, sera should be stored at 2° to 8°C
FetalClone III
Filtered using sequential 100 nm pore-size rated filters
FetalClone III Serum (U.S.)
≤ 10 EU/mL
≤ 2μg/mL
U.S.
None
≤ -10°C
USDA Inspected
Bluetongue
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit