Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ FetalClone™ III Serum (U.S.) PROMO

Designed for fibroblast applications, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ III Serum, the most widely applicable fetal bovine serum (FBS) alternative.

Manufacturer:  Cytiva 29450141

Catalog No. SH310903RR0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »

This item is not returnable. View return policy

Description

Description

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
Specifications

Specifications

500 mL
Filtered using sequential 100 nm pore-size rated filters.
FetalClone III Serum (U.S.)
≤ 10 EU/mL
Typically less than 2 µg/mL
U.S.
None
Sera should be stored at -10°C or lower. Once thawed, sera should be stored (at 2°C to 8°C.)
FetalClone III
Sterile
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Iron Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
No
≤ 20 mg/dL
No
Store at -10°C or below.
USDA Inspected
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit