Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin PROMO

A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.

$89.50 - $113.70

Specifications

Additional Assays Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Storage Requirements Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Sterility Sterile
Sterilization Method Filtered through sequential 100 nm pore-size rated filters.
View More Specs

Products 2
Catalog Number Mfr. No. Processing Price Quantity & Availability  
Catalog Number Mfr. No. Processing Price Quantity & Availability  
SH300720LR1
View Documents
Cytiva
29648036
None
Each for $89.50
Add to cart
 
SH7204HILR1
View Documents
Cytiva
29648037
Heat Inactivated
Each for $113.70
Add to cart
 
Description

Description

HyClone Iron-Supplemented Bovine Calf Serum, US Origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. After supplementation, the serum contains three to four times as much available iron and transferrin as fetal bovine serum (FBS) or equine serum.

Specifications

Specifications

Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Filtered through sequential 100 nm pore-size rated filters.
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Sterile
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit