Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare HyClone™ Iron-Supplemented Newborn Bovine Calf Serum, New Zealand Origin

A cost-effective alternative to Fetal Bovine Serum (FBS).

Manufacturer:  GE Healthcare SH30626.02IR

 View more versions of this product

Catalog No. SH3062602IR


Add to cart

Description

Description

  • Ensured safety against bovine diseases
  • Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters.
  • Ensured uniformity and consistency between bottles.
  • Specifications

    Specifications

    Sterile
    Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Saturation,Glucose,IgG
    No
    Yes
    500 mL
    Yes
    BluetongueBovine AdenovirusBovine ParvovirusBovine RSVBVDVCytopathogenic AgentsHemadsorbing AgentsRabies VirusReovirus
    USDA Regulations
    ≤ 25 EU/mL
    ≤ 25 mg/dL
    Filtered through sequential 100 nm pore-size rated filters.
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
    Irradiated Iron-Supplemented Newborn Bovine Calf Serum, New Zealand Origin
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit