Learn More
HyClone™ Minimum Essential Medium PROMO
HyClone™ Minimum Essential Medium PROMO manufactured using ISO 9001 certified processes
Manufacturer:  Cytiva 29489348
 View more versions of this product
Catalog No. SH3002401R0
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • System tested with Cytiva HyClone sera to ensure product efficacy and homogeneity
  • All products are hydrated using purified process water and undergo sterile filtration
  • Manufactured using ISO 9001 certified processes
Specifications
Specifications
0.1 μm sterile filtered
Suspension
12 Months
Liquid
2.0 mM L-glutamine, EBSS
MEM with Earle's Balanced Salt Solution (EBSS)
2° to 8°C, away from light
500 mL
cGMP, ISO 9001:2000