Learn More
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ PF-CHO™ LS and PF-CHO MPS Media
Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) SH30220.01
 View more versions of this product
Catalog No. SH3022001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
  • Specifications
    Specifications
    cGMP
    1000mL
    2° to 8°C
    HEPES
    Liquid
    PF-CHO (liquid) - Protein free
    L-glutamine, sodium bicarbonate, phenol red