Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Phosphate Buffered Saline (PBS) PROMO

Buffer cell culture growth with PBS

$4.86 - $41.25

Specifications

Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Endotoxin Concentration 1.0 EU/mL
Format Liquid
Product Type 0.067M PO4 Phosphate Buffered Saline (PBS)
Sterilization Method Filter Sterilized - 0.1 μm
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SH302560LR1
View Documents
Cytiva
29648038
500 mL
Each for $4.86
Add to cart
 
SH30256LLR1
View Documents
Cytiva
29648039
6 x 1000 mL
Pack of 6 for $41.25
Add to cart
 
Description

Description

  • QC-tested solution
Specifications

Specifications

cGMP and ISO 9001:2000 Guidelines
Liquid
Filter Sterilized - 0.1 μm
1.0 EU/mL
0.067M PO4 Phosphate Buffered Saline (PBS)
15° to 3°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit