Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ Phosphate Buffered Saline (PBS) PROMO

HyClone™ Phosphate Buffered Saline (PBS) PROMO with PBS

Specifications

Certifications/Compliance cGMP and ISO 9001:2000
Concentration 1X
Format Liquid
Product Type 0.067M Phosphate Buffered Saline (PBS)
Sterilization Method Filter Sterilized - .1 μm
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SH3025601R0
View Documents
Cytiva
29489356
500 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
SH3025602R0
View Documents
Cytiva
29489350
1 L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
SH30256FSR0
View Documents
Cytiva
29489346
6 x 500 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • QC-tested solution
Specifications

Specifications

cGMP and ISO 9001:2000
Liquid
Filter Sterilized - .1 μm
Calcium, Magnesium, Phenol Red
1X
0.067M Phosphate Buffered Saline (PBS)
3° to 15°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit