Learn More
HyClone™ SFM4CHO™ Media: PROMO
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
Manufacturer:  GE Healthcare Life Sciences SH30548.02PM
 View more versions of this product
Catalog No. SH-305-4802LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is not returnable. View return policy
Description
Description
  • Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Specifications
Specifications
cGMP
Liquid
2° to 8°C
L-Glutamine, , phenol red, HEPES
SFM4CHO media
1000mL
Sodium bicarbonate, poloxamer 188