Learn More
HyClone™ SFM4CHO™ Media: PROMO
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
Specifications
Certifications/Compliance cGMP
Storage Requirements 2° to 8°C
Description SFM4CHO media
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Format With Additives Without Additives Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Format With Additives Without Additives Price Quantity & Availability  
SH-305-4902LR View Documents GE Healthcare Life Sciences
SH30549.02PM
1000mL Liquid L-Glutamine, sodium bicarbonate, poloxamer 188 Phenol red, HEPES This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
SH-305-4802LR View Documents GE Healthcare Life Sciences
SH30548.02PM
1000mL Liquid Sodium bicarbonate, poloxamer 188 L-Glutamine, , phenol red, HEPES This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Specifications
Specifications
cGMP
2° to 8°C
SFM4CHO media