Learn More
HyClone™ SFM4Transfx 293 Media: PROMO
Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.
Manufacturer:  GE Healthcare Life Sciences SH30860.02PM
Catalog No. SH-308-6002LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is not returnable. View return policy
Description
Description
  • ADCF
  • Supports high transfection efficiency
  • Metabolically designed for high cell yield and recombinant protein/vector production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications
Specifications
cGMP
Liquid
2° to 8°C
SFM4Transfx-293 without L-Glutamine (liquid), 1000mL
1000mL
L-Glutamine; Animal Derived Component