Learn More
HyClone™ Trypsin Protease: PROMO
Derived from porcine pancreas. HyClone™ Trypsin Protease is used to disaggregate adherent cells from substrate or surface to which they are attached.
Manufacturer:  GE Healthcare Life Sciences SV30037.01PM
Catalog No. SV-300-3701LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is not returnable. View return policy
Description
Description
    • Derived from porcine pancreas
    • Gamma irradiated prior to hydration and filling
    • Formulated without calcium and magnesium
Specifications
Specifications
2.5% (10X)
Liquid
Porcine pancreas
Store frozen.
Calcium, Magnesium, EDTA, Phenol Red
Trypsin 2.5% 10X Solution, with 25.0 g Porcine Trypsin (1:250/L) in HBSS, without Calcium and Magnesium, without EDTA or Phenol Red
100mL
0.1μm sterile filtered
Porcine trypsin, HBSS, EDTA
Safety and Handling
Safety and Handling
ShelfLife : 24 months