Learn More
HyClone™ Water, Molecular Biology Grade PROMO
Click to view available options
Quantity:
6 x 1 L
For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.
  • Format: Liquid
Quantity 6 x 1 L
ShelfLife : 24 months