Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRA-POINT™ Coulometric Generator Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
125 mL
Description
- For Karl Fischer titration
Specifications
Specifications
Chemical Name or Material | HYDRA-POINT™ Coulometric Generator Solution |
Quantity | 125 mL |
Flash Point | 46.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Grade | BAKER ANALYZED™ Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction