Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydra-Point™ Coulometric Generator Solution, Universal, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
5 mL
125 mL
Packaging:
Glass Ampoule
Narrow Mouth Amber Glass Bottle
Description
- Catholyte solution for coulometric analysis. For both general use and aldehydes and ketones.
Specifications
Specifications
Flash Point | 46.00°C |
Packaging | Glass Ampoule |
Quantity | 5 mL |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Hydra-Point™ Coulometric Generator Solution, Universal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction