Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution, For Ketones and Aldehydes, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
Description
- For Coulometric Karl Fischer Analysis
Specifications
Specifications
Chemical Name or Material | HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution |
Quantity | 500 mL |
Specific Gravity | 1L = 1.10kg |
Flash Point | 48.70°C |
Name Note | For Ketones and Aldehydes |
Packaging | Narrow Mouth Amber Glass Bottle |
Grade | BAKER ANALYZED™ Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction