HYDRA-POINT™ Single Solution 5mg/mL, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer: J.T.Baker 627102

Packaging Narrow Mouth Amber Glass Bottle
Quantity 1L

 View more versions of this product

Catalog No. 02-003-107

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    Chemical Name or Material HYDRA-POINT™ Single Solution 5mg/mL
    Grade BAKER ANALYZED™ Reagent

    Specifications

    Quantity 1L
    Flash Point 46.00°C
    Packaging Narrow Mouth Amber Glass Bottle
    Chemical Name or Material HYDRA-POINT™ Single Solution 5mg/mL
    Grade BAKER ANALYZED™ Reagent

    Description

      For Karl Fischer titration

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit