Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRA-POINT™ Titrant, 5mg/mL, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Specifications
Flash Point | 26.00°C |
---|---|
Chemical Name or Material | HYDRA-POINT™ Titrant, 5mg/mL |
Packaging | Narrow Mouth Amber Glass Bottle |
Grade | BAKER ANALYZED™ Reagent |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
02-003-171
|
Avantor
627402 |
1 L | Narrow Mouth Amber Glass Bottle | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
02-003-172
|
Avantor
627468 |
2.5 L | Narrow Mouth Amber Glass Bottle | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
Description
- For Karl Fischer titration
Specifications
26.00°C | |
HYDRA-POINT™ Titrant, 5mg/mL | |
Narrow Mouth Amber Glass Bottle | |
BAKER ANALYZED™ Reagent |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title