Learn More
HYDRANAL™ - Composite 2, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Manufacturer:  Honeywell-Hydranal 348064X2.5LUS/DEL
 View more versions of this product
Catalog No. 60-046-445
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Specifications
Specifications
HYDRANAL™ - Composite 2
∼1.11 g/mL
Reagent for volumetric one-component Karl Fischer titration (methanol free)
≥70.0000%, ≥5.0000%, ≥5.0000%, ≥5.0000%, ≥5.0000%, ≥1.0000%
Glass Bottle
Karl Fischer
4 x 2.5L
103°C
111-90-0, 288-32-4, 7446-09-5, 7553-56-2, 693-98-1, 68007-08-9
<90.0000%, <10.0000%, <10.0000%, <10.0000%, <10.0000%, <5.0000%
DE Solvent, 2-(2-Ethoxyethoxy)ethanol, CARBITOL™, Diethylene glycol ethyl ether, Ethyldiglycol
Safety and Handling
Safety and Handling
ShelfLife : 1080 days from date of manufacture
Recommended Storage : Room Temp