Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™ - Composite 2, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Manufacturer: Honeywell-Hydranal 348066X1LUS/DEL
Specifications
HYDRANAL™ - Composite 2 | |
∼1.11 g/mL | |
Reagent for volumetric one-component Karl Fischer titration (methanol free) | |
≥70.0000%, ≥5.0000%, ≥5.0000%, ≥5.0000%, ≥5.0000%, ≥1.0000% | |
Glass Bottle | |
Karl Fischer |
6 x 1L | |
103°C | |
111-90-0, 288-32-4, 7446-09-5, 7553-56-2, 693-98-1, 68007-08-9 | |
<90.0000%, <10.0000%, <10.0000%, <10.0000%, <10.0000%, <5.0000% | |
DE Solvent, 2-(2-Ethoxyethoxy)ethanol, CARBITOL™, Diethylene glycol ethyl ether, Ethyldiglycol |
Safety and Handling
ShelfLife : 1080 days from date of manufacture
Recommended Storage : Room Temp
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title