Learn More
HYDRANAL™ - Coulomat AG-H, Reagent for coulometric KF titration in long-chained hydrocarbons (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™ Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Honeywell-Hydranal 348436X500MLUS
 View more versions of this product
Catalog No. 60-006-46
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
HYDRANAL™-Coulomat AG-H
11°C (51.8°F)
UN1992
500mL
67-56-1, 112-18-5, 71-41-0
Safety and Handling
Safety and Handling
P201-P210-P280-P308 + P311-P370 + P378-P403 + P235