Learn More
HYDRANAL™ - Coulomat Oil, Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™
Anolyte for coulometric Karl Fischer titration in oils (chloroform-methanol-xylene-based), preferred for cells with diaphragm
Manufacturer:  Honeywell-Hydranal 348686X100ML
 View more versions of this product
Catalog No. 60-006-51
Add to cart
Description
Description
Advantages of Hydranal Coulometric Reagents
  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
Specifications
Specifications
HYDRANAL™-Coulomat Oil
8°C (46.4°F)
UN1992
100mL
68007-08-9, 67-66-3, 95-47-6
Safety and Handling
Safety and Handling
P201-P210-P280-P308 + P311-P370 + P378-P403 + P235