Learn More
HYDRANAL™ - Coulomat Oil, Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™
Anolyte for coulometric Karl Fischer titration in oils (chloroform-methanol-xylene-based), preferred for cells with diaphragm
Supplier: Honeywell Chemicals 348686X500MLUS
Description
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)
Specifications
HYDRANAL™-Coulomat Oil | |
8°C (46.4°F) | |
UN1992 |
500 mL | |
68007-08-9, 67-66-3, 95-47-6 |
Safety and Handling
P201-P210-P280-P308 + P311-P370 + P378-P403 + P235