Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Coulomat Oil, Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™

Anolyte for coulometric Karl Fischer titration in oils (chloroform-methanol-xylene-based), preferred for cells with diaphragm

Supplier:  Honeywell Chemicals 348686X100ML/DEL

 View more versions of this product

Catalog No. 60-050-010

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

Advantages of Hydranal Coulometric Reagents

  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
Specifications

Specifications

6 x 100 mL
8°C
Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm
<50.0000%, <30.0000%, <30.0000%, <20.0000%, <5.0000%, <5.0000%
Glass Bottle
Methyl alcohol
HYDRANAL™ - Coulomat Oil
1.042 g/cm3
≥30.0000%, ≥20.0000%, ≥20.0000%, ≥10.0000%, ≥1.0000%, ≥1.0000%
95-47-6, 67-66-3, 68007-08-9, 288-32-4, 7446-09-5
UN1992
Karl Fischer
Safety and Handling

Safety and Handling

P201-P210-P280-P308 + P311-P370 + P378-P403 + P235

ShelfLife : 1800 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Flammable liquids, toxic, n.o.s.

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit