Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - KetoSolver, Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium), Honeywell Fluka™

Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free)

Manufacturer:  Honeywell-Hydranal 347386X1LUS/DEL

Catalog No. 60-046-412

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

Specifications

Specifications

HYDRANAL™ - KetoSolver
21°C
∼0.91 g/mL
107-98-2, 64-17-5, 288-32-4, 101023-55-6
<90.0000%, <20.0000%, <5.0000%, <5.0000%
UN1993
6 x 1L
85°C
Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium)
≥70.0000%, ≥10.0000%, ≥1.0000%, ≥1.0000%
Glass Bottle
Karl Fischer
Safety and Handling

Safety and Handling

ShelfLife : 1800 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Flammable liquids, n.o.s.

Recommended Storage : Room Temp

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit