Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - KetoSolver, Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium), Honeywell Fluka™
GSA_VA

Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free)

$81.90 - $120.60

Specifications

Chemical Name or Material HYDRANAL™-KetoSolver
Density 0.91g/mL
Flash Point 21°C (69.8°F)
UN Number UN1993BE
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
60-006-55
GSA_VA
View Documents
Honeywell-Hydranal
347386X500ML
500mL
Each for $81.90
Case of 6 EA for $390.00
Add to cart
 
60-006-54
View Documents
Honeywell-Hydranal
347386X1LUS
1L
Each for $120.60
Case of 6 EA for $624.00
Add to cart
 
60-046-413
View Documents
Honeywell-Hydranal
347386X500ML/DEL
6 x 500mL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
60-046-412
View Documents
Honeywell-Hydranal
347386X1LUS/DEL
6 x 1L This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

HYDRANAL™-KetoSolver
0.91g/mL
21°C (69.8°F)
UN1993BE
Safety and Handling

Safety and Handling

P201-P210-P280-P304 + P340 + P312-P308 + P313-P337 + P313

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit