Learn More
HYDRANAL™ - KetoSolver, Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium), Honeywell Fluka™
GSA_VA
Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free)
Manufacturer:  Honeywell-Hydranal 347386X500ML
 View more versions of this product
Catalog No. 60-006-55
Add to cart
Description
Description
Advantages of Hydranal one-component reagents:
  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
HYDRANAL™-KetoSolver
21°C (69.8°F)
107-98-2, 64-17-5, 288-32-4, 101023-55-6
500mL
0.91g/mL
UN1993BE
Safety and Handling
Safety and Handling
P201-P210-P280-P304 + P340 + P312-P308 + P313-P337 + P313