Learn More
HYDRANAL™ - Methanol dry, Reagent for volumetric one-component KF titration (working medium), Honeywell Fluka™
Supplier: Honeywell Chemicals 347414X2.5LUS
Description
Specifications
HYDRANAL™-Methanol dry | |
0.79g/mL | |
67-56-1 |
2.5 L | |
9°C (48.2°F) | |
UN1230 |
Safety and Handling
P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
RTECSNumber : PC1400000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov