Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Titrant 5 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Honeywell™ Fluka™

Titrating agent for volumetric two-component Karl Fischer titration (ethanol-based)

Specifications

Chemical Name or Material HYDRANAL™-Titrant 5 E
Density 0.89g/mL
Flash Point 12°C (53.6°F)
UN Number UN1170
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
60-006-80
View Documents
Honeywell-Hydranal
34732500ML
500mL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
60-006-78
View Documents
Honeywell-Hydranal
347321LUS
1L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
60-046-406
View Documents
Honeywell-Hydranal
347324X2.5L
4 x 2.5L This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
Description

Description

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

Specifications

Specifications

HYDRANAL™-Titrant 5 E
0.89g/mL
12°C (53.6°F)
UN1170
Safety and Handling

Safety and Handling

P210-P260-P305 + P351 + P338-P370 + P378-P403 + P235

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit