Learn More
Mopec Hydraulic Dissection Cart with Fiberglass X-Ray Top Available on GSA/VA Contract for Federal Government customers only.
Allow for simple elevating and tilting from either side with dual side mounted foot controls. Mopec™ Hydraulic Dissection Cart with Fiberglass X-Ray Top incorporates hydraulic lifting cylinders allowing for either end to be elevated and tilted.
Manufacturer:  Mopec DC600
Catalog No. 22-444-063
Add to cart
Description
Description
  • Twin pedestal hydraulics for easy foot activated elevation
  • Tiltable top frame
  • Four heavy duty 8 in. casters with total lock
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
1 Year Warranty
Specifications
Specifications
Mobile Autopsy Cart
32 in.
81.28cm
80.5 in.
204.47cm