Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine hydrate, 80% (Hydrazine, 51%), ACROS Organics™

Click to view available options
Quantity:
250g,250g
1kg,1kg
Packaging:
Glass bottle,Glass bottle
10217-52-4,7732-18-5
H4N2·xH2O
32.04
MFCD00149931
IKDUDTNKRLTJSI-UHFFFAOYSA-N
hydrazine hydrate,hydrazine monohydrate,hydrazinium hydroxide,hydrazine hydroxide,hydrazine, monohydrate,hydrazine, hydrate,idrazina idrata italian,hydrazinemonohydrate,unii-kyd297831p,ccris 7739
24654
CHEBI:35511
hydrazine;hydrate
NN.O
10217-52-4,7732-18-5
H4N2·xH2O
32.04
MFCD00149931
IKDUDTNKRLTJSI-UHFFFAOYSA-N
hydrazine hydrate,hydrazine monohydrate,hydrazinium hydroxide,hydrazine hydroxide,hydrazine, monohydrate,hydrazine, hydrate,idrazina idrata italian,hydrazinemonohydrate,unii-kyd297831p,ccris 7739
24654
CHEBI:35511
hydrazine;hydrate
NN.O
Reduction reagent
Color Colorless,Colorless
Boiling Point 117.2°C,117.2°C
Quantity 1kg,1kg
Packaging Glass bottle,Glass bottle
Specific Gravity 1.028,1.028
Refractive Index 1.4105 to 1.4145,1.4105 to 1.4145
Flash Point 91°C,91°C
Melting Point -57.0°C,− 57.0°C
Density 1.0280g/mL,1.0280g/mL
Linear Formula H2NNH2·xH2O
Fieser 01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174
Merck Index 14, 4771
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,insoluble in chloroform and ether
Formula Weight 32.04
Physical Form Liquid
Percent Purity 80%
Viscosity 1.33 mPa.s (20°C)
Chemical Name or Material Hydrazine hydrate
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Fatal if inhaled.
May cause cancer.
Very toxic to aquatic life with l

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plen

GHS Signal Word: Danger

RTECSNumber : MV4590000