Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Sulfate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
Packaging:
Poly Bottle
10034-93-2
H6N2O4S
130.118
ZGCHATBSUIJLRL-UHFFFAOYSA-N
hydrazine sulfate,hydrazine monosulfate,hydrazine, sulfate,hydrazine sulphate,hydrazinium sulfate,hydrazonium sulfate,hydrazine sulfate 1:1,segidrin,sehydrin,diamine sulfate
24842
hydrazine;sulfuric acid
NN.OS(=O)(=O)O
10034-93-2
H6N2O4S
130.118
ZGCHATBSUIJLRL-UHFFFAOYSA-N
hydrazine sulfate,hydrazine monosulfate,hydrazine, sulfate,hydrazine sulphate,hydrazinium sulfate,hydrazonium sulfate,hydrazine sulfate 1:1,segidrin,sehydrin,diamine sulfate
24842
hydrazine;sulfuric acid
NN.OS(=O)(=O)O

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 1.37kg
Quantity 500g
Packaging Poly Bottle
Formula Weight 130.12
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Hydrazine Sulfate