Hydrazine Sulfate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer: J.T.Baker 217704

Packaging Poly Bottle
Quantity 125g

 View more versions of this product

Catalog No. 02-003-173

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 10034-93-2
    Molecular Formula (NH2)2·H2SO4
    Formula Weight 130.12
    Chemical Name or Material Hydrazine Sulfate
    Grade Baker Analyzed™ ACS Reagent

    Specifications

    Physical Form Crystals
    Formula Weight 130.12
    Specific Gravity 1L = 1.37kg
    Molecular Formula (NH2)2·H2SO4
    Packaging Poly Bottle
    CAS 10034-93-2
    Quantity 125g
    Chemical Name or Material Hydrazine Sulfate
    Grade Baker Analyzed™ ACS Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit