Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrindantin Hydrate, Approx. 96%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
5950-69-6
C18H14O8
358.30
QHVADKNWNMILPQ-UHFFFAOYNA-N
2,2',3,3,3',3'-hexahydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1'-dione
OC1(O)C2=CC=CC=C2C(=O)C1(O)C1(O)C(=O)C2=CC=CC=C2C1(O)O
5950-69-6
C18H14O8
358.30
QHVADKNWNMILPQ-UHFFFAOYNA-N
2,2',3,3,3',3'-hexahydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1'-dione
OC1(O)C2=CC=CC=C2C(=O)C1(O)C1(O)C(=O)C2=CC=CC=C2C1(O)O

H2451, 5950-69-6, C18H10O6•2H2O

Spectrum™ Hydrindantin Hydrate
Packaging Amber Glass Bottle
Quantity 100 g
CAS Max % 1
Percent Purity Approx. 96%