Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrion™ pH Test Papers, Wide-Range
Select the pH range you need, quickly and inexpensively.
Supplier: Micro Essential Lab 160B/EMD
Description
- Type A indicates pH 2, 4, 6, 8 and 10
- Type B indicates pH 1, 3, 5, 7, 9 and 11
Order Info
- Cat. No. S60146 and S60148: 5 rolls and 2 color charts per kit, 10 kits per case
- Cat. No. S60152: 10 vials per box
Specifications
1 to 11 | |
Color comparison scale | |
Refill Roll | |
4.57 m |
To determine pH within half unit via distinct color changes | |
5 Pack | |
15 ft. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction