Learn More
PicoStudio Hydro Turbine
SCI_ED
The Hydro Turbine explores how turbines are designed and built, and how they generate energy.
Manufacturer:  PicoStudio PS30042
 View more versions of this product
Catalog No. S4131931
Add to cart
Alert:
Small Parts - Not recommended for children under 3 years old.
Description
Description
  • Explore the power of water energy with our Hydro Turbine!
  • Learn how turbines are designed and built, and how they generate energy.
  • Construct this STEM kit from scratch and engineer it to power an LED, piezo buzzer, and showcase its power on a standard multimeter!
  • Check out our other levels to create endless experiments with your turbine!
Specifications
Specifications
14 in.
Hydro Level 1 Parts, 1 Long Tower, 1 Weighted Base Insert, 1 AC Generator Pack (1 Coil Housing, 1 AC Shaft, 1 Magnet Clip Set, 2 O-Rings, 1 4-Count Coil Pack, 1 4-Count Magnet Pack), 1 Rectification Lab (1 Solderless Breadboard, 1 Piezo Buzzer, 8 Male End Wires, 2 Capacitors, 4 Diodes)
228.6 mm
Level 2
355.6 mm
9 in.
STEM Kit