Learn More
PicoStudio Hydro Turbine Products for Science Education
The Hydro Turbine explores how turbines are designed and built, and how they generate energy.
$34.99 - $109.99
Specifications
Product Type STEM Kit
View More Specs
Alert:
Small Parts - Not recommended for children under 3 years old.
Products 3
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
S4131930 Products for Science Education
View Documents
PicoStudio
PS30041
Level 1 1 Base, 1 Short Tower 139.7 mm (5.5 in.), 1 Nacelle Base, 1 Nacelle Front, 1 Nose Cone, 2 Blade Hubs, 6 Blade Adapters, 6 Hydro Paddles, 1 DC Shaft, 1 DC Generator Pack (1 DC Motor, 1 DC Motor Stand), 1 Alligator Clip Set (Black and Red), 1 Screw Pack (1 LED, 1 Piezo Buzzer, 6 Screws, 6 Nuts, 1 Resistor 30 Ohm, 1 Motor Clip)
Each for $34.99
Add to cart
S4131931 Products for Science Education
View Documents
PicoStudio
PS30042
Level 2 Hydro Level 1 Parts, 1 Long Tower, 1 Weighted Base Insert, 1 AC Generator Pack (1 Coil Housing, 1 AC Shaft, 1 Magnet Clip Set, 2 O-Rings, 1 4-Count Coil Pack, 1 4-Count Magnet Pack), 1 Rectification Lab (1 Solderless Breadboard, 1 Piezo Buzzer, 8 Male End Wires, 2 Capacitors, 4 Diodes)
Each for $79.99
Add to cart
S4131932 Products for Science Education
View Documents
PicoStudio
PS30043
Level 3 Hydro Level 2 Parts, 1 AC Generator Pack, 1 Rectification Lab, 1 Long Tower, 1 Weighted Base Insert, 1 Circuit 101 Lab (3 Resistors 10, 30, 100 Ohm, 2 AA Battery Holder, 1 Light bulb 1.5V, 8 Male End Wires, 1 Light Bulb Stand, 1 Alligator Clips Set (Red and Black), 1 Multimeter), 1 Solar Panel 7.2V 200mA
Each for $109.99
Add to cart
Description
Description
  • Explore the power of water energy with our Hydro Turbine!
  • Learn how turbines are designed and built, and how they generate energy.
  • Construct this STEM kit from scratch and engineer it to power an LED, piezo buzzer, and showcase its power on a standard multimeter!
  • Check out our other levels to create endless experiments with your turbine!
Specifications
Specifications
STEM Kit