Learn More
MCR Safety Hydroblast™ 3 Goggle
Click to view available options
Lens Tint:
Clear
Gray
Description
- Low profile athletic design
- 7-Point Ratchet action temple for all day comfort and adjustability
- Dielectric
- Appropriate for use in chemical environments
Specifications
Lens Tint | Clear |
Frame Color | Blue |
Ventilation Details | Indirect vent |