Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, 33% Solution in Acetic Acid, Technical, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
100 kg
Packaging:
Amber Glass Bottle
Drum
10035-10-6,64-19-7
BrH
80.91
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide
[H+].[Br-]
10035-10-6,64-19-7
BrH
80.91
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide
[H+].[Br-]

H1205, 10035-10-6

Spectrum™ Hydrobromic Acid, 33 Percent Solution in Acetic Acid, Technical a mineral acid and one of the strongest known. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability
Quantity 100 kg
CAS Max % 0.33,0.67
UN Number UN2920
Packaging Drum