Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, 47% to 49%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

10035-10-6
BrH
80.912
CPELXLSAUQHCOX-UHFFFAOYSA-N
260
CHEBI:47266
bromane
Br

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Quantity 500mL
Formula Weight 80.92
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.50kg
Chemical Name or Material Hydrobromic Acid
Assay 47 to 49%
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements

Recommended Storage : Material darkens when exposed to air and light