Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, 47-49% (Certified ACS), Fisher Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass Bottle
10035-10-6,7732-18-5
BrH
80.912
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide
260
CHEBI:47266
bromane
Br
10035-10-6,7732-18-5
BrH
80.912
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide
260
CHEBI:47266
bromane
Br

Note: This reagent may darken on standing

Color Yellow
Quantity 500mL
pH <1
Assay Percent Range 47.0 to 49.0 %
UN Number 1788
Chloride 0.05% max.
Density 1.480g/cm³
Formula Weight 80.91
Physical Form Liquid
Percent Purity 47.0 to 49.0%
Grade Certified ACS
Heavy Metals (as Pb) 5ppm max.
Identification Pass Test
Ignition Residue 0.002% max.
Iron (Fe) 1ppm max.
Iodide 0.003% max.
Packaging Glass Bottle
Phosphate 0.001% max.
Selenium (Se) 0.01ppm max.
Sulfate and Sulfite 0.003% max.
Vapor Pressure 9 mmHg at 25°C
Melting Point -11°C
Boiling Point 126°C to 128°C
Chemical Name or Material Hydrobromic Acid, 47-49%
Show More Show Less

DOTInformation : DOT Class 8, : Corrosive