Learn More
Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals
Description
Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Appearance | Colorless to pale yellow liquid |
Color | Colorless to Yellow |
Density | 1.49 |
Sensitivity | Air Sensitive |
Vapor Pressure | 22500 hPa (16876mm Hg) |
Assay Percent Range | 99.9999% (metals basis) |
Quantity | 5 mL |
Physical Form | Liquid |
Beilstein | 3587158 |
Solubility Information | Miscible with water. |
Show More |
Safety and Handling
Hazard Category | H290-H314-H335 |
Hazard Statement | Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. |
Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
DOTInformation | Hazard Class: 8, Packing Group: II |
EINECSNumber | 233-113-0 |
RTECSNumber | MW3850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.